missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tri(2-thienyl)phosphine, TCI America™

Manufacturer:  TCI America T16661G

 View more versions of this product

Catalog No. T16661G



Specifications

Specifications

Tri(2-thienyl)phosphine
Undesignated
24171-89-9
MFCD00015471
KUCPTMZJPDVWJL-UHFFFAOYSA-N
trithiophen-2-ylphosphane
90384
Crystal-Lump at 20°C
205°C
1g
C12H9PS3
tris 2-thienyl phosphine, phosphine, tri-2-thienyl, tri 2-thienyl phosphine, tri thiophen-2-yl phosphine, tri2-thienylphosphine, tris thiophen-2-yl phosphane, tri-2-thienylphosphine, trithienylphosphine, pubchem6427, tri 2-thienyl phosphine #
C1=CSC(=C1)P(C2=CC=CS2)C3=CC=CS3
280.358
280.36
≥96.0% (GC)

Chemical Identifiers

24171-89-9
280.358
KUCPTMZJPDVWJL-UHFFFAOYSA-N
90384
C1=CSC(=C1)P(C2=CC=CS2)C3=CC=CS3
C12H9PS3
MFCD00015471
tris 2-thienyl phosphine, phosphine, tri-2-thienyl, tri 2-thienyl phosphine, tri thiophen-2-yl phosphine, tri2-thienylphosphine, tris thiophen-2-yl phosphane, tri-2-thienylphosphine, trithienylphosphine, pubchem6427, tri 2-thienyl phosphine #
trithiophen-2-ylphosphane
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents