Please login to your online account to display your discounted pricing

tri(2-methylphenyl)phosphine, Maybridge

Chemical Identifiers

Chemical Name or Material Tri(2-methylphenyl)phosphine
Assay 97%
Assay Percent Range 97%
Molecular Formula C21H21P
Linear Formula (CH3C6H4)3P
Formula Weight 304.37
Synonym Tri(2-methylphenyl)phosphine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MB00147EE

  SDS
fisher scientific company
MB00147EE
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 304.37
Quantity 50g
Assay 97%
CAS 6163-58-2
Molecular Formula C21H21P
Synonym Tri(2-methylphenyl)phosphine
Assay Percent Range 97%
Chemical Name or Material Tri(2-methylphenyl)phosphine
Health Hazard 2 GHS P Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula (CH3C6H4)3P