missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tri(2-methylphenyl)phosphine, Maybridge

Manufacturer:  Thermo Scientific MB00147EE

 View more versions of this product

Catalog No. MB00147EE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

50g
6163-58-2
C21H21P
phosphine tris 2-methylphenyl, tris-2-methylphenyl phosphane, tris 2-methylphenyl phosphine, tris 2-methyl-phenyl phosphine, tris-2-methylphenyl phosphine, tris 2-methylphenyl-phosphine, phosphine tris methylphenyl, tri o-tolyl phosphine 1g, tri-2-methylphenyl phosphine
CC1=CC=CC=C1P(C2=CC=CC=C2C)C3=CC=CC=C3C
304.373
304.37
Tri(2-methylphenyl)phosphine
97%
(CH3C6H4)3P
COIOYMYWGDAQPM-UHFFFAOYSA-N
tris(2-methylphenyl)phosphane
80271
97%

Chemical Identifiers

6163-58-2
304.373
phosphine tris 2-methylphenyl, tris-2-methylphenyl phosphane, tris 2-methylphenyl phosphine, tris 2-methyl-phenyl phosphine, tris-2-methylphenyl phosphine, tris 2-methylphenyl-phosphine, phosphine tris methylphenyl, tri o-tolyl phosphine 1g, tri-2-methylphenyl phosphine
tris(2-methylphenyl)phosphane
C21H21P
COIOYMYWGDAQPM-UHFFFAOYSA-N
80271
CC1=CC=CC=C1P(C2=CC=CC=C2C)C3=CC=CC=C3C
Safety and Handling

Safety and Handling

GHS P Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents