missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tri-(2-furyl)phosphine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 294802500

 View more versions of this product

Catalog No. AC294802500



Specifications

Specifications

Tri-(2-furyl)phosphine
136.0°C (4.0 mmHg)
250mg
5518-52-5
C12H9O3P
tri 2-furyl phosphine, tri fur-2-yl phosphane, tri furan-2-yl phosphane, tri furan-2-yl phosphine, tri-2-furyl phosphine, tri-2-furylphosphine, trifurylphosphine, tris 2-furanyl phosphine, tris 2-furyl phosphine, tris furan-2-yl phosphane
C1=COC(=C1)P(C2=CC=CO2)C3=CC=CO3
232.18
232.18
Glass bottle
61.0°C to 64.0°C
98%
98%
MFCD00151857
DLQYXUGCCKQSRJ-UHFFFAOYSA-N
tris(furan-2-yl)phosphane
521585
98%

Chemical Identifiers

5518-52-5
232.18
DLQYXUGCCKQSRJ-UHFFFAOYSA-N
521585
C1=COC(=C1)P(C2=CC=CO2)C3=CC=CO3
C12H9O3P
MFCD00151857
tri 2-furyl phosphine, tri fur-2-yl phosphane, tri furan-2-yl phosphane, tri furan-2-yl phosphine, tri-2-furyl phosphine, tri-2-furylphosphine, trifurylphosphine, tris 2-furanyl phosphine, tris 2-furyl phosphine, tris furan-2-yl phosphane
tris(furan-2-yl)phosphane
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS Signal Word: Warning

Recommended Storage : Refrigerator +4°C

SDS Documents