Please login to your online account to display your discounted pricing

Tri(1-naphthyl)phosphine, +96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tri(1-naphthyl)phosphine
CAS 3411-48-1
Assay ≥96.0%
Assay Percent Range 96+%
Molecular Formula C30H21P
Formula Weight 412.47
MDL Number MFCD00046400
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC390160050

  SDS
thermo fisher scientific
AC390160050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC390160010

  SDS
thermo fisher scientific
AC390160010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder
Formula Weight 412.47
Quantity 5g
Color Slightly yellow
Infrared Spectrum Authentic
CAS 3411-48-1
MDL Number MFCD00046400
Molecular Formula C30H21P
Chemical Name or Material Tri(1-naphthyl)phosphine
Packaging Glass bottle
Assay Percent Range 96+%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.