missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri(1-aziridinyl)phosphine sulfide, 98%

Manufacturer:  Alfa Aesar™ H6177803

 View more versions of this product

Catalog No. AAH6177803



Description

Description

Solubility
Soluble in benzene, acetone and methanol.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

Tri(1-aziridinyl)phosphine sulfide
52-24-4
MFCD00145452
tespamin
C1CN1P(=S)(N2CC2)N3CC3
189.217
CHEBI:9570
Solid
1g
C6H12N3PS
UN2811
FOCVUCIESVLUNU-UHFFFAOYSA-N
tris(aziridin-1-yl)-sulfanylidene-$l^{5}-phosphane
5453
189.22
98%

Chemical Identifiers

52-24-4
189.217
FOCVUCIESVLUNU-UHFFFAOYSA-N
5453
tris(aziridin-1-yl)-sulfanylidene-$l^{5}-phosphane
C6H12N3PS
MFCD00145452
tespamin
CHEBI:9570
C1CN1P(=S)(N2CC2)N3CC3
Safety and Handling

Safety and Handling

GHS H Statement
H300-H340-H350
Fatal if swallowed.
May cause genetic defects.
May cause cancer.

GHS P Statement
P281-P301+P310-P308+P313-P321-P405-P501a
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF exposed or concerned: Get medical advice/attention.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : SZ2975000

storageNote1 : 2° to 8°C

Documents