missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri(1-aziridinyl)phosphine sulfide, 98%

$165.01 - $758.73

Chemical Identifiers

CAS 52-24-4
Molecular Formula C6H12N3PS
Molecular Weight (g/mol) 189.217
MDL Number MFCD00145452
InChI Key FOCVUCIESVLUNU-UHFFFAOYSA-N
Synonym tespamin
PubChem CID 5453
ChEBI CHEBI:9570
IUPAC Name tris(aziridin-1-yl)-sulfanylidene-$l^{5}-phosphane
SMILES C1CN1P(=S)(N2CC2)N3CC3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6177803 View Documents Alfa Aesar™
H6177803
1g Each for $165.01
 
AAH6177806 View Documents Alfa Aesar™
H6177806
5g Each for $758.73
 
Description

Description

Solubility
Soluble in benzene, acetone and methanol.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

52-24-4
189.217
FOCVUCIESVLUNU-UHFFFAOYSA-N
5453
tris(aziridin-1-yl)-sulfanylidene-$l^{5}-phosphane
C6H12N3PS
MFCD00145452
tespamin
CHEBI:9570
C1CN1P(=S)(N2CC2)N3CC3

Specifications

1g
C6H12N3PS
UN2811
FOCVUCIESVLUNU-UHFFFAOYSA-N
tris(aziridin-1-yl)-sulfanylidene-$l^{5}-phosphane
5453
189.22
98%
52-24-4
MFCD00145452
tespamin
C1CN1P(=S)(N2CC2)N3CC3
189.217
CHEBI:9570
Solid
Tri(1-aziridinyl)phosphine sulfide
Safety and Handling

Safety and Handling

GHS H Statement
H300-H340-H350
Fatal if swallowed.
May cause genetic defects.
May cause cancer.

GHS P Statement
P281-P301+P310-P308+P313-P321-P405-P501a
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF exposed or concerned: Get medical advice/attention.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : SZ2975000

storageNote1 : 2° to 8°C