missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Trazodone hydrochloride

A serotonin uptake inhibitor

$84.96 - $345.43

Chemical Identifiers

CAS 25332-39-2
Molecular Formula C19H23Cl2N5O
Molecular Weight (g/mol) 408.327
MDL Number MFCD00079603
InChI Key OHHDIOKRWWOXMT-UHFFFAOYSA-N
Synonym trazodone hydrochloride, trazodone hcl, desyrel, bimaran, molipaxin, devidon, pragmazone, thombran, tombran, tritico
PubChem CID 62935
ChEBI CHEBI:9655
IUPAC Name 2-[3-[4-(3-chlorophenyl)piperazin-1-yl]propyl]-[1,2,4]triazolo[4,3-a]pyridin-3-one;hydrochloride
SMILES C1CN(CCN1CCCN2C(=O)N3C=CC=CC3=N2)C4=CC(=CC=C4)Cl.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6307003 View Documents Alfa Aesar™
J6307003
1g Each for $84.96
 
AAJ6307006 View Documents Alfa Aesar™
J6307006
5g Each for $345.43
 
Specifications

Chemical Identifiers

25332-39-2
408.327
OHHDIOKRWWOXMT-UHFFFAOYSA-N
62935
2-[3-[4-(3-chlorophenyl)piperazin-1-yl]propyl]-[1,2,4]triazolo[4,3-a]pyridin-3-one;hydrochloride
C19H23Cl2N5O
MFCD00079603
trazodone hydrochloride, trazodone hcl, desyrel, bimaran, molipaxin, devidon, pragmazone, thombran, tombran, tritico
CHEBI:9655
C1CN(CCN1CCCN2C(=O)N3C=CC=CC3=N2)C4=CC(=CC=C4)Cl.Cl

Specifications

White
1g
C19H23Cl2N5O
14,9579
Soluble at 25mg/ml in methanol; Also soluble in 0.1M HCl or DMSO; Insoluble in water
C1CN(CCN1CCCN2C(=O)N3C=CC=CC3=N2)C4=CC(=CC=C4)Cl.Cl
408.327
CHEBI:9655
Powder
222°C to 227°C
25332-39-2
MFCD00079603
trazodone hydrochloride, trazodone hcl, desyrel, bimaran, molipaxin, devidon, pragmazone, thombran, tombran, tritico
OHHDIOKRWWOXMT-UHFFFAOYSA-N
2-[3-[4-(3-chlorophenyl)piperazin-1-yl]propyl]-[1,2,4]triazolo[4,3-a]pyridin-3-one;hydrochloride
62935
408.33
Trazodone hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H351-H302
Suspected of causing cancer.
Harmful if swallowed.

GHS P Statement
P281-P264-P301+P312-P308+P313-P405-P501a
Use personal protective equipment as required.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : XZ5660000