Learn More
Thermo Scientific™ Trap Column Cartridges Holders with nanoViper Fittings
Works with virtually any valve and any column from any manufacturer.
Manufacturer: Thermo Scientific™ 164649
Description
NanoViper connections come preassembled and high pressure tested. Thermo Scientific Trap Column Cartridges Holders with nanoViper fittings do not "grab" the tubing at a single point. They seal at the tip without using a ferrule. This gives nanoViper fittings their UHPLC, ensuing virtually dead-volume-free capabilities.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
nanoLC | |
Trap Column Cartridge Holder | |
5 mm | |
30 µm (fitting) |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.