CORNING™ TransportoCells™ Vector Control
CORNING™ TransportoCells™ Vector Control are suitable for use as negative control for SLC transporter-expressing TransportoCells™
Supplier: CORNING™ 354854
Description
- Single use, cryopreserved mammalian HEK-293 cells transfected with vector DNA
- Use as negative control for SLC transporter-expressing TransportoCells
- One vial contains enough for one 24-well or one 96-well plate
Specifications
−135°C to −200°C | |
Human | |
1/Pk, ≥10 Million cells | |
Each vial is sufficient for one 24-well or one 96-well plate |
293 (HEK) | |
Negative control for SLC transporter-expressing TransportoCells | |
Human |