missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
CORNING™ TransportoCells™ Human Organic Anion-Transporting Polypeptide 1B1*1a; Wild Type (388A)
CORNING™ TransportoCells™ Human Organic Anion-Transporting Polypeptide 1B1*1a; Wild Type (388A)
Supplier: CORNING™ 354859
Description
- Single use, cryopreserved mammalian HEK-293 cells transiently overexpressing Human OATP1B1*1a protein
- Robust and consistent performance with uptake ratios ≥, sufficient for one 24-well or one 96-well plate
Specifications
−135°C to −200°C | |
Human | |
Human |
293 (HEK) | |
1/Pk, ≥10 Million cells | |
Transporter: OATP1B1*1a/SLCO1B1*1a, each vial is sufficient for one 24-well or one 96-well plate |