missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ TransportoCell™ ABC Vesicle
Designed for rapid compound screening. TransportoCell™ ABC Vesicle is a mamalian expression system used for pre screening assay.
$1,458.54 - $2,197.10
Specifications
Content And Storage | -80°C |
---|---|
Quantity | 0.5 mL |
Sample Volume | 0.5 mL |
Stability | 60 months |
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity | ||||
CG453803
|
Corning™
453803 |
Human MRP2 Vesicle | 0.5 mL |
Each for $2,197.10
|
|
CG453802
|
Corning™
453802 |
Human BSEP Vesicle | 0.5 mL |
Each for $2,197.10
|
|
CG453804
|
Corning™
453804 |
Human BCRP Vesicle | 0.5 mL |
Each for $2,197.10
|
|
CG453800
|
Corning™
453800 |
Vesicle Negative Control | 0.5 mL |
Each for $1,458.54
|
|
CG453801
|
Corning™
453801 |
Human MDR1 Vesicle | 0.5 mL |
Each for $2,197.10
|
|
Description
- Mammalian cell expression system
- High throughput models
- Ideal for use in pre screening assays
- Measures drug candidates ability
- Can be used with non radiolabelled isotopes
- Measurres drug stimulated vanadate-sensitive ATPase activity
- Mammalian cell expression system
- Higher uptake activity
- Lower background
- Eliminates biosafety concern from baculovirus
- Improve in vitro to in vivo prediction
Specifications
-80°C | |
0.5 mL | |
0.5 mL | |
60 months |