missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Transition Metals, plasma standard solution, Specpure™, Cd, Co, Cr, Cu, Fe, Mn, Ni, V, Zn at 100μg/mL

Manufacturer:  Alfa Aesar™ 44518AE

 View more versions of this product

Catalog No. AA44518AE



Specifications

Specifications

Transition Metals, plasma standard solution
MFCD00151266
Liquid
100mL
Cd, Co, Cr, Cu, Fe, Mn, Ni, V, Zn at 100μg/mL
UN3264
Specpure™
Matrix: 5% HNO3
Documents