Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Transglutaminase II Ab-4, Rabbit Polyclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Transglutaminase II Ab-4, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB060P1ABX

 View more versions of this product

Catalog No. RB060P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Regulatory Status RUO
Clone Polyclonal
Host Species Rabbit
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG
Format Concentrated
Antigen Transglutaminase II Ab-4
Quantity 500µL
Immunogen Recombinant full length human TGase II
Formulation Ab without BSA/Azide (Frozen)
Conjugate Unlabeled
Type Primary
Classification Polyclonal
Research Discipline Neuroscience
Concentration 1mg/mL
Item Description 500uL; 1mg/mL; Unlabeled; Purified without BSA and azide

Provide accurate, reproducible results with the Thermo Scientific™ Transglutaminase II Ab-4, Rabbit Polyclonal Antibody.

TGase II is implicated in programmed cell death, signal transduction, drug-resistance, cell growth, endocytosis, insulin secretion, cell adhesion, cataract formation, and wound healing.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant full-length human TGase II

Molecular Weight: 77-85kDa

Positive Control: HUVEC cells, Breast CA

Cellular Localization: Cytoplasm, membrane and extracellular matrix Ab-4 is excellent for staining of endothelial cells in murine tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO