Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Transglutaminase II Ab-3, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Transglutaminase II Ab-3, Mouse Monoclonal Antibody.

$300.16 - $1,487.31

Specifications

Antigen Transglutaminase II Ab-3
Clone CUB 7402 + TG100
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS300P

 
thermo scientific™ lab vision™
MS300P
Purified Ab with BSA and Azide 1mL Each for $1,487.31

MS300P0

 
thermo scientific™ lab vision™
MS300P0
Purified Ab with BSA and Azide 100µL Each for $300.16

MS300P1ABX

 
thermo scientific™ lab vision™
MS300P1ABX
Ab without BSA/Azide (Frozen) 100µL Each for $850.35

MS300PABX

 
thermo scientific™ lab vision™
MS300PABX
Ab without BSA/Azide (Frozen) 200µL Each for $1,431.37

MS300R7

 
thermo scientific™ lab vision™
MS300R7
Ready-to-use for IHC (UVLP) 7mL Each for $721.02
Description & Specifications

Specifications

Antigen Transglutaminase II Ab-3
Clone CUB 7402 + TG100
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Guinea Pig
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline Neuroscience
Type Primary
Immunogen Purified transglutaminase II from guinea pig liver
Isotype IgG1

Provide accurate, reproducible results with the Thermo Scientific™ Transglutaminase II Ab-3, Mouse Monoclonal Antibody.

Transglutaminase II catalyzes calcium-dependent post-translational modification of proteins by formation of an isopeptide bond within or between polypeptide chains. It is also known as TGC, tTG, type II-, Gh, cytosolic-, liver-, endothelial-, erythrocyte-, cellular-transglutaminase. Different tissues and cell types express varying amounts of tissue transglutaminase with a markedly hugh expression in rheumatoid lesions. It is implicated in programmed cell death, signal transduction, drug-resistance, cell growth, endocytosis, insulin secretion, cell adhesion, cataract formation, and wound healing.

Host Species: Mouse

Clone: CUB 7402 + TG100

Isotype: IgG1

Species Reactivity: Human and Guinea Pig. Others not known.

Immunogen: Purified transglutaminase II from guinea pig liver

Molecular Weight: 77-85kDa

Positive Control: HUVEC cells, Breast CA

Cellular Localization: Cytoplasm, membrane and extracellular matrix

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO