Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Transglutaminase II Ab-3, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Transglutaminase II Ab-3, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS300P1ABX

 View more versions of this product

Catalog No. MS300P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Transglutaminase II Ab-3
Format Concentrated
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Purified transglutaminase II from guinea pig liver
Isotype IgG1
Regulatory Status RUO
Cross Reactivity Guinea Pig
Cross Reactivity Human
Host Species Mouse
Quantity 100µL
Clone CUB 7402 + TG100
Formulation Ab without BSA/Azide (Frozen)
Type Primary
Concentration 1mg/mL
Research Discipline Neuroscience
Classification Monoclonal
Conjugate Unlabeled
Item Description 100uL; 1mg/mL; Purified w/o BSA, azide

Provide accurate, reproducible results with the Thermo Scientific™ Transglutaminase II Ab-3, Mouse Monoclonal Antibody.

Transglutaminase II catalyzes calcium-dependent post-translational modification of proteins by formation of an isopeptide bond within or between polypeptide chains. It is also known as TGC, tTG, type II-, Gh, cytosolic-, liver-, endothelial-, erythrocyte-, cellular-transglutaminase. Different tissues and cell types express varying amounts of tissue transglutaminase with a markedly hugh expression in rheumatoid lesions. It is implicated in programmed cell death, signal transduction, drug-resistance, cell growth, endocytosis, insulin secretion, cell adhesion, cataract formation, and wound healing.

Host Species: Mouse

Clone: CUB 7402 + TG100

Isotype: IgG1

Species Reactivity: Human and Guinea Pig. Others not known.

Immunogen: Purified transglutaminase II from guinea pig liver

Molecular Weight: 77-85kDa

Positive Control: HUVEC cells, Breast CA

Cellular Localization: Cytoplasm, membrane and extracellular matrix

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO