Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Transglutaminase II Ab-2, Mouse Monoclonal Antibody

Provides accurate, reproducible results with the Thermo Scientific™ Transglutaminase II Ab-2, Mouse Monoclonal Antibody.

$259.39 - $1,532.20

Specifications

Antigen Transglutaminase II Ab-2
Clone TG100
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS279B

 
thermo scientific™ lab vision™
MS279B
Biotinylated Ab with BSA and Azide 1mL Each for $1,532.20

MS279B0

 
thermo scientific™ lab vision™
MS279B0
Biotinylated Ab with BSA and Azide 100µL Each for $354.96

MS279B1

 
thermo scientific™ lab vision™
MS279B1
Biotinylated Ab with BSA and Azide 500µL Each for $955.90

MS279P

 
thermo scientific™ lab vision™
MS279P
Purified Ab with BSA and Azide 1mL Each for $1,220.53

MS279P0

 
thermo scientific™ lab vision™
MS279P0
Purified Ab with BSA and Azide 100µL Each for $259.39

MS279P1

 
thermo scientific™ lab vision™
MS279P1
Purified Ab with BSA and Azide 500µL Each for $742.04

MS279P1ABX

 
thermo scientific™ lab vision™
MS279P1ABX
Ab without BSA/Azide (Frozen) 100µL Each for $825.33

MS279PABX

 
thermo scientific™ lab vision™
MS279PABX
Ab without BSA/Azide (Frozen) 200µL Each for $1,323.78

MS279R7

 
thermo scientific™ lab vision™
MS279R7
Ready-to-use for IHC (UVLP) 7mL Each for $696.69
Description & Specifications

Specifications

Antigen Transglutaminase II Ab-2
Clone TG100
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Rat
Host Species Mouse
Regulatory Status RUO
Research Discipline Neuroscience
Type Primary
Immunogen Purified guinea pig liver TGase II
Isotype IgG1

Provides accurate, reproducible results with the Thermo Scientific™ Transglutaminase II Ab-2, Mouse Monoclonal Antibody.

Transglutaminase II catalyzes calcium-dependent post-translational modification of proteins by formation of an isopeptide bond within or between polypeptide chains. It is also known as TGC, tTG, type II-, Gh, cytosolic-, liver-, endothelial-, erythrocyte-, cellular-transglutaminase. Different tissues and cell types express varying amounts of tissue transglutaminase with a markedly hugh expression in rheumatoid lesions. It is implicated in programmed cell death, signal transduction, drug-resistance, cell growth, endocytosis, insulin secretion, cell adhesion, cataract formation, and wound healing.

Host Species: Mouse

Clone: TG100

Isotype: IgG1

Species Reactivity: Human, Rat and Guinea Pig. Others not known.

Epitope: aa 447-538

Immunogen: Purified guinea pig liver TGase II

Molecular Weight: 77-85kDa

Positive Control: HUVEC cells, Breast CA

Cellular Localization: Cytoplasm, membrane and extracellular matrix Ab-2 is the Ab-of-Choice as a marker for endothelial cells in rat tissues

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO