missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Transforming Growth Factor α (TGFα) Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Transforming Growth Factor α (TGFα) Ab-1, Mouse Monoclonal Antibody.

$337.88 - $1,779.96

Specifications

Antigen Transforming Growth Factor alpha (TGFalpha) Ab-1
Clone MF9
Applications Western Blot
Classification Monoclonal
Format Concentrated
View More Specs

Products 8
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity  
MS670B View Documents Thermo Scientific™
MS670B
200μg/mL Biotin 1mL Each for $1,779.96
 
MS670B0 View Documents Thermo Scientific™
MS670B0
200μg/mL Biotin 100μL Each for $452.16
 
MS670B1 View Documents Thermo Scientific™
MS670B1
200μg/mL Biotin 500μL Each for $1,192.10
 
MS670P View Documents Thermo Scientific™
MS670P
200μg/mL Unconjugated 1mL Each for $1,367.55
 
MS670P0 View Documents Thermo Scientific™
MS670P0
200μg/mL Unconjugated 100μL Each for $337.88
 
MS670P1 View Documents Thermo Scientific™
MS670P1
200μg/mL Unconjugated 500μL Each for $911.43
 
MS670P1ABX View Documents Thermo Scientific™
MS670P1ABX
1mg/mL Unconjugated 100μL Each for $1,012.82
 
MS670PABX View Documents Thermo Scientific™
MS670PABX
1mg/mL Unconjugated 200μL Each for $1,472.96
 
Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Transforming Growth Factor α (TGFα) Ab-1, Mouse Monoclonal Antibody.

TGFα (aa 50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.

Host Species: Mouse

Clone: MF9

Isotype: IgG1/κ

Species Reactivity: Human, Cow, Rat and Mouse. Others not known.

Epitope: C-terminal

Immunogen: A 17-amino acid synthetic peptide from the carboxyl-terminus of rat TGFα

Molecular Weight: 6kDa

Positive Control: Recombinant TGFα protein, A2058 cells

Cellular Localization: Cytoplasmic Ab-1 shows no cross-reaction with EGF and the neuropeptide synenkephalin. Ab-1 is excellent for staining of formalin-fixed, paraffin-embedded tissues. The staining with Ab-1 is completely blocked by the peptide used for raising this antibody.

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Transforming Growth Factor alpha (TGFalpha) Ab-1
Western Blot
Concentrated
RUO
Primary
IgG2a kappa
MF9
Monoclonal
Murine
Signal Transduction
A 17-amino acid synthetic peptide from the carboxyl-terminus of Rat TGFalpha