Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Transforming Growth Factor α (TGFα) Ab-1, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Transforming Growth Factor α (TGFα) Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS670P1ABX

 View more versions of this product

Catalog No. MS670P1ABX

  • / Each
 

Description & Specifications

Specifications

Immunogen A 17-amino acid synthetic peptide from the carboxyl-terminus of Rat TGFalpha
Antigen Transforming Growth Factor alpha (TGFalpha) Ab-1
Format Concentrated
Applications Western Blotting
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Clone MF9
Quantity 100µL
Host Species Mouse
Isotype IgG2a kappa
Regulatory Status RUO
Research Discipline Signal Transduction
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled

Provide accurate, reproducible results with the Thermo Scientific™ Transforming Growth Factor α (TGFα) Ab-1, Mouse Monoclonal Antibody.

TGFα (aa 50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.

Host Species: Mouse

Clone: MF9

Isotype: IgG1/κ

Species Reactivity: Human, Cow, Rat and Mouse. Others not known.

Epitope: C-terminal

Immunogen: A 17-amino acid synthetic peptide from the carboxyl-terminus of rat TGFα

Molecular Weight: 6kDa

Positive Control: Recombinant TGFα protein, A2058 cells

Cellular Localization: Cytoplasmic Ab-1 shows no cross-reaction with EGF and the neuropeptide synenkephalin. Ab-1 is excellent for staining of formalin-fixed, paraffin-embedded tissues. The staining with Ab-1 is completely blocked by the peptide used for raising this antibody.

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO