Please login to your online account to display your discounted pricing

Alfa Aesar™ Transformation buffer-1, pH 7.5

Chemical Identifiers

Chemical Name or Material Transformation buffer-1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ63244EQE

  SDS
alfa aesar™
J63244EQE
125mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Transformation buffer-1
pH 7.5
Quantity 125mL
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Color Colorless
Physical Form Liquid
Recommended Storage 2° to 8°C

Applications
Transformation buffer is used for transformation of competent cells.

Solubility
Difficult to mix.

Notes
Contains 30mM potassium-acetate, 150mM calcium chloride, 10mM PIPES, 100mM potassium chloride, 50mM manganese chloride and 15% glycerol, pH 7.5

Safety and Handling

Warning

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H319
Causes serious eye irritation.

2° to 8°C