BrandTech™ Transferpette™ S Single Channel Adjustable Pipet
Exibits a high degree of accuracy, precision, maximum versatility, and reliable performance with Easy Calibration™ technology. BrandTech™ Transferpette™ S Single Channel Adjustable Pipet offers simple one-handed operation with an adjustable brake to protect against accidental volume changes.
Manufacturer:  BrandTech™ 704773
 View more versions of this product
Catalog No. 03-837-330
Includes: Shelf/rack mount, performance certificate, operating manual
Description
Description
The Transferpette™ S Single Channel Adjustable Pipet offers robust yet lightweight construction, simple, one-handed operation, a high degree of accuracy and precision with Easy Calibration™ technology for long lasting reliability. The single channel model is available in fixed volume and digital models in volume ranges from 0.1µL to 10mL.
  • Centralized pipetting button with optimized stroke length
  • Adjustable brake to fix volume range
  • Visible four-digit volume display
  • Compatible with universal tips (0.1-1μL requires nano-cap tips)
  • Repeatedly autoclavable at 121°C/250°F(20min.)
Warranty
Three-year warranty
Specifications
Specifications
±0.8%
CE, IVD
Digital
Demanding laboratory applications
Performance certificate, operating manual, shelf mount, and three year warranty.
3 Years
Single channel pipet
Yes
2 to 200µL tip, 5 to 300µL tip, 2 to 20µL tip, 5 to 100µL tip
Curved
2 to 200µL tip, 5 to 300µL tip, 2 to 20µL tip, 5 to 100µL tip
0.05μL
D-50
5 to 50μL
Documents