Promega Transcription Factor Consensus Oligonucleotides DFS Item
These oligonucleotides contain consensus DNA-binding sites for individual transcription factors and have 5′ OH blunt ends, allowing labeling to high specific activity with T4 PNK.
Manufacturer:  Promega E3231
 View more versions of this product
Catalog No. PR-E3231
Description
Description
  • Oligo Containing DNA-Binding Site for a Transcription Factor
  • Designed with 5′ OH blunt ends; easily labeled to high specific activity with T4 polynucleotide kinase
  • Supplied at a concentration of 1.75pM/µL
Specifications
Specifications
1.75 pmol/μL
SP1 Consensus Oligonucleotide
Store at -20°C
Gel-shift assays, Affinity chromatography
175 pmol