Please login to your online account to display your discounted pricing

Alfa Aesar™ trans,trans,trans-1,5,9-Cyclododecatriene, 97%, stab. with 30-50ppm 4-tert-butylcatechol

Chemical Identifiers

Chemical Name or Material trans,trans,trans-1,5,9-Cyclododecatriene
CAS 676-22-2
Name Note stabilized with 30 to 50ppm 4-tert-butylcatechol
Molecular Formula C12H18
Formula Weight 162.28
MDL Number MFCD00003720
UN Number UN1759
Beilstein 969880
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0931706

  SDS
alfa aesar™
L0931706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0931714

  SDS
alfa aesar™
L0931714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 162.28
MDL Number MFCD00003720
Chemical Name or Material trans,trans,trans-1,5,9-Cyclododecatriene
CAS 676-22-2
Assay 97%
Molecular Formula C12H18
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Health Hazard 1 Danger
Odor Pungent
Beilstein 969880
Flash Point 73°C (163°F)
Name Note stabilized with 30 to 50ppm 4-tert-butylcatechol
Melting Point 32° to 34°C
Quantity 5g
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
Boiling Point 237° to 238°C
UN Number UN1759
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.