missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans,trans-Muconic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC128070050

 View more versions of this product

Catalog No. AC128070050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

trans, trans-Muconic acid
Authentic
≥96.0%
C6H6O4
142.11
02, 803
15, 6385
Beige
3588-17-8
Crystalline Powder
97%
HO2CCH=CHCH=CHCO2H
MFCD00002702
Glass bottle
trans, trans-1,3-Butadiene-1,4-dicarboxylic acid
5g

Chemical Identifiers

97%
HO2CCH=CHCH=CHCO2H
MFCD00002702
trans, trans-Muconic acid
C6H6O4
142.11
trans, trans-1,3-Butadiene-1,4-dicarboxylic acid
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.