missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans,trans-Dibenzylideneacetone, 98+%

Manufacturer:  Alfa Aesar™ A1209514

 View more versions of this product

Catalog No. AAA1209514



Description

Description

Applications
trans,trans-Dibenzylideneacetone was used as an additive in the copper-catalyzed-arylation of imidazoles. It is a reactant involved in Nazarov-like cyclization, Transfer hydrogenation, Lewis acid mediated condensation, Hetero-Diels-Alder reactions, Asymmetric 1,4-addition reactions and Michael addition reactions.

Solubility
Soluble in chloroform, acetone, alcohol (slightly), and hot methanol. Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications

Specifications

trans,trans-Dibenzylideneacetone
25g
C17H14O
608434
dibenzylideneacetone, 1,5-diphenylpenta-1,4-dien-3-one, trans,trans-dibenzylideneacetone, dibenzalacetone, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one, dibenzylidene acetone, distyryl ketone, 1,5-diphenyl-1,4-pentadien-3-one, trans,trans-dibenzalacetone, trans,trans-1,5-diphenyl-1,4-pentadien-3-one
C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2
234.298
234.3
112°C to 114°C
35225-79-7
MFCD00004790
14,3006
WMKGGPCROCCUDY-PHEQNACWSA-N
(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one
640180
≥98%

Chemical Identifiers

35225-79-7
234.298
WMKGGPCROCCUDY-PHEQNACWSA-N
640180
C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2
C17H14O
MFCD00004790
dibenzylideneacetone, 1,5-diphenylpenta-1,4-dien-3-one, trans,trans-dibenzylideneacetone, dibenzalacetone, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one, dibenzylidene acetone, distyryl ketone, 1,5-diphenyl-1,4-pentadien-3-one, trans,trans-dibenzalacetone, trans,trans-1,5-diphenyl-1,4-pentadien-3-one
(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one
SDS Documents