Please login to your online account to display your discounted pricing

Alfa Aesar™ trans,trans,cis-1,5,9-Cyclododecatriene, 98+%

Chemical Identifiers

Chemical Name or Material trans,trans,cis-1,5,9-Cyclododecatriene
CAS 706-31-0
Molecular Formula C12H18
Formula Weight 162.28
MDL Number MFCD00063609
UN Number UN2518
Beilstein 1362442
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB204180B

  SDS
alfa aesar™
B204180B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2041830

  SDS
alfa aesar™
B2041830
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 162.28
Chemical Name or Material trans,trans,cis-1,5,9-Cyclododecatriene
TSCA TSCA
Assay ≥98%
Molecular Formula C12H18
MDL Number MFCD00063609
CAS 706-31-0
UN Number UN2518
Beilstein 1362442
Melting Point −18°C
Boiling Point 230° to 232°C
Health Hazard 1 Danger
Quantity 1000g
Flash Point 73°C (163°F)
Health Hazard 2 GHS H Statement
H331-H227-H315-H319-H317-H335
Toxic if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 1,5,9-CYCLODODECATRIENE
RTECS Number GU2310000

Applications
trans,trans,cis-1,5,9-Cyclododecatriene, is used as a precursor in the production of nylon-12. It is also used as a chemical intermediate (butadiene trimerisation), precursor for dodecanedioic acid (antiseptics, coatings, corrosion inhibitor, surfactant, engineering plastics; PA12X), laurolactam (precursor to the polyamide PA12).

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H227-H315-H319-H317-H335
Toxic if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.