missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans,trans-4′-Propyl-4-(3,4,5-trifluorophenyl)bicyclohexyl, TCI America™

Manufacturer:  TCI America P231425G

 View more versions of this product

Catalog No. P231425G



Specifications

Specifications

trans,trans-4′-Propyl-4-(3,4,5-trifluorophenyl)bicyclohexyl
66°C
131819-23-3
trans,trans-4'-propyl-4-3,4,5-trifluorophenyl bicyclohexyl, trans-4-3,4,5-trifluorophenyl-trans-4'-propylbicyclohexane, trans-4-n-propyl-4'-3,4,5-trifluorophenyl bicyclohexyl, benzene, 1,2,3-trifluoro-5-trans,trans-4'-propyl 1,1'-bicyclohexyl-4-yl, 4-3,4,5-trifluorophenyl-4'-propyl-1,1'-bicyclohexane, trans,trans-1-4'-propylbicyclohexyl-3,4,5-trifluorobenzene, 1,2,3-trifluoro-5-trans,trans-4-propyl 1,1-bicyclohexyl-4-yl benzene, all-trans-4-propyl-4'-3,4,5-trifluorophenyl bi cyclohexane, 1s,1'r,4r,4'r-4-propyl-4'-3,4,5-trifluorophenyl-1,1'-bi cyclohexane, 1s,4r-4-propyl-4'-3,4,5-trifluorophenyl-1,1'-bi cyclohexane
CCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C(=C3)F)F)F
338.458
338.46
≥98.0% (GC)
White
25g
C21H29F3
FEWMLRARKGRCCE-UHFFFAOYSA-N
1,2,3-trifluoro-5-[4-(4-propylcyclohexyl)cyclohexyl]benzene
11772113
Crystal-Powder at 20°C

Chemical Identifiers

131819-23-3
338.458
trans,trans-4'-propyl-4-3,4,5-trifluorophenyl bicyclohexyl, trans-4-3,4,5-trifluorophenyl-trans-4'-propylbicyclohexane, trans-4-n-propyl-4'-3,4,5-trifluorophenyl bicyclohexyl, benzene, 1,2,3-trifluoro-5-trans,trans-4'-propyl 1,1'-bicyclohexyl-4-yl, 4-3,4,5-trifluorophenyl-4'-propyl-1,1'-bicyclohexane, trans,trans-1-4'-propylbicyclohexyl-3,4,5-trifluorobenzene, 1,2,3-trifluoro-5-trans,trans-4-propyl 1,1-bicyclohexyl-4-yl benzene, all-trans-4-propyl-4'-3,4,5-trifluorophenyl bi cyclohexane, 1s,1'r,4r,4'r-4-propyl-4'-3,4,5-trifluorophenyl-1,1'-bi cyclohexane, 1s,4r-4-propyl-4'-3,4,5-trifluorophenyl-1,1'-bi cyclohexane
1,2,3-trifluoro-5-[4-(4-propylcyclohexyl)cyclohexyl]benzene
C21H29F3
FEWMLRARKGRCCE-UHFFFAOYSA-N
11772113
CCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C(=C3)F)F)F
Safety and Handling

Safety and Handling

EINECSNumber : (4)-1877

TSCA : No

Documents