missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans,trans-4'-n-Propylbicyclohexyl-4-carboxylic acid, 97%

$65.43 - $284.43

Chemical Identifiers

CAS 65355-32-0
Molecular Formula C16H28O2
Molecular Weight (g/mol) 252.398
MDL Number MFCD05864407
InChI Key JXPGQFKJNKWDKP-UHFFFAOYSA-N
Synonym 1,1'-bicyclohexyl-4-carboxylic acid, 4'-propyl
PubChem CID 793726
IUPAC Name 4-(4-propylcyclohexyl)cyclohexane-1-carboxylic acid
SMILES CCCC1CCC(CC1)C2CCC(CC2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5527106 View Documents Alfa Aesar™
H5527106
5g Each for $65.43
 
AAH5527114 View Documents Alfa Aesar™
H5527114
25g Each for $284.43
 
Specifications

Chemical Identifiers

65355-32-0
252.398
JXPGQFKJNKWDKP-UHFFFAOYSA-N
793726
CCCC1CCC(CC1)C2CCC(CC2)C(=O)O
C16H28O2
MFCD05864407
1,1'-bicyclohexyl-4-carboxylic acid, 4'-propyl
4-(4-propylcyclohexyl)cyclohexane-1-carboxylic acid

Specifications

200°C
65355-32-0
MFCD05864407
JXPGQFKJNKWDKP-UHFFFAOYSA-N
4-(4-propylcyclohexyl)cyclohexane-1-carboxylic acid
793726
97%
5g
C16H28O2
1,1'-bicyclohexyl-4-carboxylic acid, 4'-propyl
CCCC1CCC(CC1)C2CCC(CC2)C(=O)O
252.398
252.4
trans,trans-4'-n-Propylbicyclohexyl-4-carboxylic acid
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures