missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
trans,trans-4'-n-Propylbicyclohexyl-4-carboxylic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H5527106
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
trans,trans-4'-n-Propylbicyclohexyl-4-carboxylic acid | |
200°C | |
MFCD05864407 | |
trans-4'-propyl-1,1'-bicyclohexyl-4-carboxylic acid, 1,1'-bicyclohexyl-4-carboxylic acid, 4'-propyl, 4-4-propylcyclohexyl cyclohexanecarboxylic acid, 4-4-propylcyclohexyl cyclohexane-1-carboxylic acid, trans-4'-propyl-1,1'-bicyclohexyl-4-carboxylicacid, jxpgqfkjnkwdkp-ktslabgisa-n, 4'-propylbicyclohexane-4-carboxylic acid, 4-propyl-4'-bicyclohexanecarboxylic acid, trans-4-propylbis cyclohexane carboxylic acid, 4'-propyl-1,1'-bicyclohexyl-4-carboxylic acid | |
CCCC1CCC(CC1)C2CCC(CC2)C(=O)O | |
252.398 | |
252.4 |
65355-32-0 | |
C16H28O2 | |
5 g | |
JXPGQFKJNKWDKP-UHFFFAOYSA-N | |
4-(4-propylcyclohexyl)cyclohexane-1-carboxylic acid | |
793726 | |
97% |
Chemical Identifiers
65355-32-0 | |
252.398 | |
JXPGQFKJNKWDKP-UHFFFAOYSA-N | |
793726 | |
CCCC1CCC(CC1)C2CCC(CC2)C(=O)O |
C16H28O2 | |
MFCD05864407 | |
trans-4'-propyl-1,1'-bicyclohexyl-4-carboxylic acid, 1,1'-bicyclohexyl-4-carboxylic acid, 4'-propyl, 4-4-propylcyclohexyl cyclohexanecarboxylic acid, 4-4-propylcyclohexyl cyclohexane-1-carboxylic acid, trans-4'-propyl-1,1'-bicyclohexyl-4-carboxylicacid, jxpgqfkjnkwdkp-ktslabgisa-n, 4'-propylbicyclohexane-4-carboxylic acid, 4-propyl-4'-bicyclohexanecarboxylic acid, trans-4-propylbis cyclohexane carboxylic acid, 4'-propyl-1,1'-bicyclohexyl-4-carboxylic acid | |
4-(4-propylcyclohexyl)cyclohexane-1-carboxylic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures