missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
trans,trans-4'-Ethyl-4-(3,4,5-trifluorophenyl)bicyclohexyl 98.0+%, TCI America™
Supplier: TCI America E115625G
Specifications
trans,trans-4′-Ethyl-4-(3,4,5-trifluorophenyl)bicyclohexyl | |
73°C | |
C20H27F3 | |
25 g | |
DXFHMMVZUZLQFU-UHFFFAOYSA-N | |
5-[4-(4-ethylcyclohexyl)cyclohexyl]-1,2,3-trifluorobenzene | |
14984828 | |
≥98.0% (GC) |
139215-80-8 | |
White | |
MFCD12911825 | |
1r,4r-4-ethyl-4'-3,4,5-trifluorophenyl-1,1'-bi cyclohexane, benzene, 5-trans,trans-4'-ethyl 1,1'-bicyclohexyl-4-yl-1,2,3-trifluoro, 4-ethyl-4'-3,4,5-trifluorophenyl bi cyclohexane, trans-4-ethyl-4'-3,4,5-trifluorophenyl bicyclohexyl, 4-ethyl-4'-3,4,5-trifluorophenyl-1,1'-bi cyclohexane, 5-4-4-ethylcyclohexyl cyclohexyl-1,2,3-trifluorobenzene, acmc-20myma, 4-ethyl-4'-3,4,5-trifluorophenyl-1,1'-bi cyclohexyl, trans,trans-4-4'-ethylbicyclohexyl-1,2,3-trifluorobenzene | |
CCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C(=C3)F)F)F | |
324.431 | |
324.43 | |
Crystalline Powder |
Chemical Identifiers
139215-80-8 | |
324.431 | |
DXFHMMVZUZLQFU-UHFFFAOYSA-N | |
14984828 | |
CCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C(=C3)F)F)F |
C20H27F3 | |
MFCD12911825 | |
1r,4r-4-ethyl-4'-3,4,5-trifluorophenyl-1,1'-bi cyclohexane, benzene, 5-trans,trans-4'-ethyl 1,1'-bicyclohexyl-4-yl-1,2,3-trifluoro, 4-ethyl-4'-3,4,5-trifluorophenyl bi cyclohexane, trans-4-ethyl-4'-3,4,5-trifluorophenyl bicyclohexyl, 4-ethyl-4'-3,4,5-trifluorophenyl-1,1'-bi cyclohexane, 5-4-4-ethylcyclohexyl cyclohexyl-1,2,3-trifluorobenzene, acmc-20myma, 4-ethyl-4'-3,4,5-trifluorophenyl-1,1'-bi cyclohexyl, trans,trans-4-4'-ethylbicyclohexyl-1,2,3-trifluorobenzene | |
5-[4-(4-ethylcyclohexyl)cyclohexyl]-1,2,3-trifluorobenzene |
Safety and Handling
EINECSNumber : (2)-3938
TSCA : No