missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans,trans-4′-Butyl-4-(3,4-difluorophenyl)bicyclohexyl, TCI America™

$259.73 - $898.36

Chemical Identifiers

CAS 82832-58-4
Molecular Formula C22H32F2
Molecular Weight (g/mol) 334.495
MDL Number MFCD13188634
InChI Key NZXZINXFUSKTPH-UHFFFAOYSA-N
Synonym trans,trans-4-butyl-4'-3,4-difluorophenyl-1,1'-bi cyclohexane, trans,trans-4-3,4-difluorophenyl-4-butylbicyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-butyl-bicyclohexyl, 4-4-4-butylcyclohexyl cyclohexyl-1,2-difluorobenzene, 4-trans,trans-4'-butyl 1,1'-bicyclohexyl-4-yl-1,2-difluorobenzene, 4-butyl-4'-3,4-difluorophenyl-1,1'-bi cyclohexane, 1s,4r-4-butyl-4'-3,4-difluorophenyl-1,1'-bi cyclohexane, 1s,1'r,4r,4'r-4-butyl-4'-3,4-difluorophenyl-1,1'-bi cyclohexane, trans,trans-4'-butyl-4-3,4-difluorophenyl bicyclohexyl, trans,trans-4-butyl-4-3,4-difluorophenyl bicyclohexyl
PubChem CID 14993439
IUPAC Name 4-[4-(4-butylcyclohexyl)cyclohexyl]-1,2-difluorobenzene
SMILES CCCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B49255G View Documents TCI America
B49255G
5g Each for $259.73
 
B492525G View Documents TCI America
B492525G
25g Each for $898.36
 
Specifications

Chemical Identifiers

82832-58-4
334.495
NZXZINXFUSKTPH-UHFFFAOYSA-N
14993439
CCCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F
C22H32F2
MFCD13188634
trans,trans-4-butyl-4'-3,4-difluorophenyl-1,1'-bi cyclohexane, trans,trans-4-3,4-difluorophenyl-4-butylbicyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-butyl-bicyclohexyl, 4-4-4-butylcyclohexyl cyclohexyl-1,2-difluorobenzene, 4-trans,trans-4'-butyl 1,1'-bicyclohexyl-4-yl-1,2-difluorobenzene, 4-butyl-4'-3,4-difluorophenyl-1,1'-bi cyclohexane, 1s,4r-4-butyl-4'-3,4-difluorophenyl-1,1'-bi cyclohexane, 1s,1'r,4r,4'r-4-butyl-4'-3,4-difluorophenyl-1,1'-bi cyclohexane, trans,trans-4'-butyl-4-3,4-difluorophenyl bicyclohexyl, trans,trans-4-butyl-4-3,4-difluorophenyl bicyclohexyl
4-[4-(4-butylcyclohexyl)cyclohexyl]-1,2-difluorobenzene

Specifications

White
5g
C22H32F2
trans,trans-4-butyl-4'-3,4-difluorophenyl-1,1'-bi cyclohexane, trans,trans-4-3,4-difluorophenyl-4-butylbicyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-butyl-bicyclohexyl, 4-4-4-butylcyclohexyl cyclohexyl-1,2-difluorobenzene, 4-trans,trans-4'-butyl 1,1'-bicyclohexyl-4-yl-1,2-difluorobenzene, 4-butyl-4'-3,4-difluorophenyl-1,1'-bi cyclohexane, 1s,4r-4-butyl-4'-3,4-difluorophenyl-1,1'-bi cyclohexane, 1s,1'r,4r,4'r-4-butyl-4'-3,4-difluorophenyl-1,1'-bi cyclohexane, trans,trans-4'-butyl-4-3,4-difluorophenyl bicyclohexyl, trans,trans-4-butyl-4-3,4-difluorophenyl bicyclohexyl
CCCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F
334.495
334.49
≥98.0% (GC)
47°C
82832-58-4
MFCD13188634
NZXZINXFUSKTPH-UHFFFAOYSA-N
4-[4-(4-butylcyclohexyl)cyclohexyl]-1,2-difluorobenzene
14993439
Crystal-Powder at 20°C
trans,trans-4′-Butyl-4-(3,4-difluorophenyl)bicyclohexyl
Safety and Handling

Safety and Handling

TSCA : No