missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5535606
Description
It is  used in liquid crystal monomer, pharmaceutical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is  used in liquid crystal monomer, pharmaceutical synthesis.
Solubility
Slightly soluble in water.
Notes
Incompatible with oxidizing agents. Store at room temperature.
Specifications
trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl | |
43°C to 46°C | |
MFCD09838997 | |
trans,trans-4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 1,2-difluoro-4-4-4-propylcyclohexyl cyclohexyl benzene, trans,trans-4-3,4-difluorophenyl-4-propylbicyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-propyl-bicyclohexyl, 4-3,4-difluoro-phenyl-4'-propyl-bicyclohexyl, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-n-propylbicyclohexyl, benzene, 1,2-difluoro-4-trans,trans-4'-propyl 1,1'-bicyclohexyl-4-yl, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 4-3,4-difluorophenyl-4'-propyl-bicyclohexyl | |
FSWZOZXLWVWJAH-UHFFFAOYSA-N | |
1,2-difluoro-4-[4-(4-propylcyclohexyl)cyclohexyl]benzene | |
581325 | |
97% |
82832-57-3 | |
C21H30F2 | |
5 g | |
Slightly soluble in water. | |
CCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F | |
320.468 | |
320.47 |
Chemical Identifiers
82832-57-3 | |
320.468 | |
FSWZOZXLWVWJAH-UHFFFAOYSA-N | |
581325 | |
CCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F |
C21H30F2 | |
MFCD09838997 | |
trans,trans-4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 1,2-difluoro-4-4-4-propylcyclohexyl cyclohexyl benzene, trans,trans-4-3,4-difluorophenyl-4-propylbicyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-propyl-bicyclohexyl, 4-3,4-difluoro-phenyl-4'-propyl-bicyclohexyl, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-n-propylbicyclohexyl, benzene, 1,2-difluoro-4-trans,trans-4'-propyl 1,1'-bicyclohexyl-4-yl, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 4-3,4-difluorophenyl-4'-propyl-bicyclohexyl | |
1,2-difluoro-4-[4-(4-propylcyclohexyl)cyclohexyl]benzene |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only