missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl, 97%

Manufacturer:  Alfa Aesar™ H5535603

 View more versions of this product

Catalog No. AAH5535603



Description

Description

Applications
It is  used in liquid crystal monomer, pharmaceutical synthesis.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents. Store at room temperature.
Specifications

Specifications

trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl
1g
C21H30F2
1,2-difluoro-4-4-4-propylcyclohexyl cyclohexyl benzene, 4-3,4-difluoro-phenyl-4'-propyl-bicyclohexyl, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexyl, 4-3,4-difluorophenyl-4'-propyl-bicyclohexyl, benzene, 1,2-difluoro-4-trans,trans-4'-propyl 1,1'-bicyclohexyl-4-yl, trans,trans-4-3,4-difluorophenyl-4'-n-propylbicyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, trans,trans-4-3,4-difluorophenyl-4'-propyl-bicyclohexyl, trans,trans-4-3,4-difluorophenyl-4-propylbicyclohexyl
FSWZOZXLWVWJAH-UHFFFAOYSA-N
1,2-difluoro-4-[4-(4-propylcyclohexyl)cyclohexyl]benzene
581325
97%
43°C to 46°C
82832-57-3
MFCD09838997
Slightly soluble in water.
CCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F
320.468
320.47

Chemical Identifiers

82832-57-3
320.468
FSWZOZXLWVWJAH-UHFFFAOYSA-N
581325
CCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F
C21H30F2
MFCD09838997
1,2-difluoro-4-4-4-propylcyclohexyl cyclohexyl benzene, 4-3,4-difluoro-phenyl-4'-propyl-bicyclohexyl, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexyl, 4-3,4-difluorophenyl-4'-propyl-bicyclohexyl, benzene, 1,2-difluoro-4-trans,trans-4'-propyl 1,1'-bicyclohexyl-4-yl, trans,trans-4-3,4-difluorophenyl-4'-n-propylbicyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, trans,trans-4-3,4-difluorophenyl-4'-propyl-bicyclohexyl, trans,trans-4-3,4-difluorophenyl-4-propylbicyclohexyl
1,2-difluoro-4-[4-(4-propylcyclohexyl)cyclohexyl]benzene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents