missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
trans,trans-4-(3,4-Difluorophenyl)-4'-n-pentylbicyclohexyl, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5516803
Description
It is used as flavor and fragrance intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as flavor and fragrance intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Slightly soluble in water.
Notes
Reacts with strong oxidizing agents. Keep at room temperature.
Specifications
trans,trans-4-(3,4-Difluorophenyl)-4'-n-pentylbicyclohexyl | |
45°C to 47°C | |
MFCD13182310 | |
trans,trans-4-3,4-difluorophenyl-4'-pentyl-1,1'-bi cyclohexane, trans,trans-4-3,4-difluorophenyl-4'-pentylbicyclohexyl, 4-trans-4-trans-4-pentylcyclohexyl cyclohexyl-1,2-difluorobenzene, benzene, 1,2-difluoro-4-4'-pentyl 1,1'-bicyclohexyl-4-yl, 1,2-difluoro-4-trans,trans-4'-pentyl 1,1'-bicyclohexyl-4-yl benzene, 1,2-difluoro-4-4-4-pentylcyclohexyl cyclohexyl benzene, 4-3,4-difluorophenyl-4'-pentyl-1,1'-bi cyclohexane, trans,trans-4-3,4-difluorophenyl-4'-n-pentylbicyclohexyl, 1r,1's,4r,4'r-4-3,4-difluorophenyl-4'-pentyl-1,1'-bi cyclohexane, trans,trans-4-pentylbicyclohexyl-3,4-difluorobenzene | |
YDVUSMRUBCJGAV-UHFFFAOYSA-N | |
1,2-difluoro-4-[4-(4-pentylcyclohexyl)cyclohexyl]benzene | |
581521 | |
97% |
118164-51-5 | |
C23H34F2 | |
1 g | |
Slightly soluble in water. | |
CCCCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F | |
348.522 | |
348.52 |
Chemical Identifiers
118164-51-5 | |
348.522 | |
YDVUSMRUBCJGAV-UHFFFAOYSA-N | |
581521 | |
CCCCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F |
C23H34F2 | |
MFCD13182310 | |
trans,trans-4-3,4-difluorophenyl-4'-pentyl-1,1'-bi cyclohexane, trans,trans-4-3,4-difluorophenyl-4'-pentylbicyclohexyl, 4-trans-4-trans-4-pentylcyclohexyl cyclohexyl-1,2-difluorobenzene, benzene, 1,2-difluoro-4-4'-pentyl 1,1'-bicyclohexyl-4-yl, 1,2-difluoro-4-trans,trans-4'-pentyl 1,1'-bicyclohexyl-4-yl benzene, 1,2-difluoro-4-4-4-pentylcyclohexyl cyclohexyl benzene, 4-3,4-difluorophenyl-4'-pentyl-1,1'-bi cyclohexane, trans,trans-4-3,4-difluorophenyl-4'-n-pentylbicyclohexyl, 1r,1's,4r,4'r-4-3,4-difluorophenyl-4'-pentyl-1,1'-bi cyclohexane, trans,trans-4-pentylbicyclohexyl-3,4-difluorobenzene | |
1,2-difluoro-4-[4-(4-pentylcyclohexyl)cyclohexyl]benzene |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only