missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans,trans-2,4-Nonadienal, TCI America™

$91.67 - $273.48

Chemical Identifiers

CAS 5910-87-2
Molecular Formula C9H14O
Molecular Weight (g/mol) 138.21
MDL Number MFCD00007006
InChI Key ZHHYXNZJDGDGPJ-BSWSSELBSA-N
Synonym 2,4-nonadienal, trans,trans-2,4-nonadienal, 2e,4e-nona-2,4-dienal, nonadienal, 2,4-nonadienal, e,e, 2,4-nonadienal, 2e,4e, 2,4-trans,trans-nonadienal, trans,trans-nona-2,4-dienal, unii-y411g77sh7, trans,trans-2,4-nonadien-1-al
PubChem CID 5283339
IUPAC Name (2E,4E)-nona-2,4-dienal
SMILES CCCCC=CC=CC=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N052125ML View Documents TCI America
N052125ML
25mL Each for $273.48
 
N05215ML View Documents TCI America
N05215ML
5mL Each for $91.67
 
Specifications

Chemical Identifiers

5910-87-2
138.21
ZHHYXNZJDGDGPJ-BSWSSELBSA-N
5283339
CCCCC=CC=CC=O
C9H14O
MFCD00007006
2,4-nonadienal, trans,trans-2,4-nonadienal, 2e,4e-nona-2,4-dienal, nonadienal, 2,4-nonadienal, e,e, 2,4-nonadienal, 2e,4e, 2,4-trans,trans-nonadienal, trans,trans-nona-2,4-dienal, unii-y411g77sh7, trans,trans-2,4-nonadien-1-al
(2E,4E)-nona-2,4-dienal

Specifications

99°C
25mL
C9H14O
2,4-nonadienal, trans,trans-2,4-nonadienal, 2e,4e-nona-2,4-dienal, nonadienal, 2,4-nonadienal, e,e, 2,4-nonadienal, 2e,4e, 2,4-trans,trans-nonadienal, trans,trans-nona-2,4-dienal, unii-y411g77sh7, trans,trans-2,4-nonadien-1-al
CCCCC=CC=CC=O
138.21
138.21
≥85.0% (GC)
Yellow
5910-87-2
MFCD00007006
ZHHYXNZJDGDGPJ-BSWSSELBSA-N
(2E,4E)-nona-2,4-dienal
5283339
Clear Liquid at 20°C
trans,trans-2,4-Nonadienal
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2462

TSCA : Yes