missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans,trans-2,4-Nonadienal 85.0+%, TCI America™

Manufacturer:  TCI America N052125ML

 View more versions of this product

Catalog No. N052125ML



Specifications

Specifications

trans,trans-2,4-Nonadienal
Yellow
5910-87-2
MFCD00007006
ZHHYXNZJDGDGPJ-BSWSSELBSA-N
(2E,4E)-nona-2,4-dienal
5283339
Clear Liquid at 20°C
99°C
25mL
C9H14O
2,4-nonadienal, trans,trans-2,4-nonadienal, 2e,4e-nona-2,4-dienal, nonadienal, 2,4-nonadienal, e,e, 2,4-nonadienal, 2e,4e, 2,4-trans,trans-nonadienal, trans,trans-nona-2,4-dienal, unii-y411g77sh7, trans,trans-2,4-nonadien-1-al
CCCCC=CC=CC=O
138.21
138.21
≥85.0% (GC)

Chemical Identifiers

5910-87-2
138.21
ZHHYXNZJDGDGPJ-BSWSSELBSA-N
5283339
CCCCC=CC=CC=O
C9H14O
MFCD00007006
2,4-nonadienal, trans,trans-2,4-nonadienal, 2e,4e-nona-2,4-dienal, nonadienal, 2,4-nonadienal, e,e, 2,4-nonadienal, 2e,4e, 2,4-trans,trans-nonadienal, trans,trans-nona-2,4-dienal, unii-y411g77sh7, trans,trans-2,4-nonadien-1-al
(2E,4E)-nona-2,4-dienal
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2462

TSCA : Yes

Documents