Please login to your online account to display your discounted pricing

Alfa Aesar™ trans,trans-2,4-Hexadienyl acetate, 97%

$36.31 - $104.42

Chemical Identifiers

Chemical Name or Material trans,trans-2,4-Hexadienyl acetate
CAS 1516-17-2
Molecular Formula C8H12O2
Formula Weight 140.18
MDL Number MFCD00036521
Beilstein 1702607
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1105006

  SDS
alfa aesar™
L1105006
5g Each for $36.31

AAL1105014

  SDS
alfa aesar™
L1105014
25g Each for $104.42
Description & Specifications

Specifications

Formula Weight 140.18
Chemical Name or Material trans,trans-2,4-Hexadienyl acetate
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P310-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Beilstein 1702607
Health Hazard 1 Danger
MDL Number MFCD00036521
TSCA TSCA
Molecular Formula C8H12O2
Health Hazard 2 GHS H Statement
H318-H227
Causes serious eye damage.
Combustible liquid.
Quantity 5g
Flash Point 71°C (159°F)
Boiling Point 100°C (20mmHg)
RTECS Number AI0350000
CAS 1516-17-2

Applications
trans,trans-2,4-Hexadienyl acetate is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Safety and Handling

Danger

GHS P Statement
P210-P280-P305+P351+P338-P310-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H227
Causes serious eye damage.
Combustible liquid.