missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans,trans-2,4-Hexadienyl acetate, 97%

Manufacturer:  Alfa Aesar™ L1105006

 View more versions of this product

Catalog No. AAL1105006



Description

Description

Applications
trans,trans-2,4-Hexadienyl acetate is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Specifications

trans,trans-2,4-Hexadienyl acetate
100°C (20mmHg)
1516-17-2
MFCD00036521
sorbyl acetate, 2,4-hexadienyl acetate, trans,trans-2,4-hexadienyl acetate, hexa-2,4-dienyl acetate, 2,4-hexadien-1-ol, acetate, unii-5416sj1t8t, acetic acid, 2,4-hexadien-1-ol ester, acetic acid, 2,4-hexadienyl ester, 2,4-hexadien-1-ol, 1-acetate, 2,4-hexadienylester kyseliny octove
CC=CC=CCOC(=O)C
140.182
140.18
71°C (159°F)
5g
C8H12O2
1702607
PXVKYPFROMBALG-VNKDHWASSA-N
[(2E,4E)-hexa-2,4-dienyl] acetate
5363491
97%

Chemical Identifiers

1516-17-2
140.182
PXVKYPFROMBALG-VNKDHWASSA-N
5363491
CC=CC=CCOC(=O)C
C8H12O2
MFCD00036521
sorbyl acetate, 2,4-hexadienyl acetate, trans,trans-2,4-hexadienyl acetate, hexa-2,4-dienyl acetate, 2,4-hexadien-1-ol, acetate, unii-5416sj1t8t, acetic acid, 2,4-hexadien-1-ol ester, acetic acid, 2,4-hexadienyl ester, 2,4-hexadien-1-ol, 1-acetate, 2,4-hexadienylester kyseliny octove
[(2E,4E)-hexa-2,4-dienyl] acetate
Safety and Handling

Safety and Handling

GHS H Statement
H318-H227
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P280-P305+P351+P338-P310-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : AI0350000

TSCA : TSCA

Documents