Please login to your online account to display your discounted pricing

Alfa Aesar™ trans,trans-2,4-Hexadien-1-ol, 98%, stab. with 0.1% alpha-tocopherol

$37.85 - $418.69

Chemical Identifiers

Chemical Name or Material trans,trans-2,4-Hexadien-1-ol
CAS 17102-64-6
Name Note stabilized with 0.1% alpha-tocopherol
Molecular Formula C6H10O
Formula Weight 98.15
MDL Number MFCD00002925
Beilstein 1719718
Synonym 1-Hydroxy-2,4-hexadiene; Sorbyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1266006

 
alfa aesar™
A1266006
5g Each for $37.85

AAA1266014

 
alfa aesar™
A1266014
25g Each for $120.69

AAA1266022

 
alfa aesar™
A1266022
100g Each for $418.69
Description & Specifications

Specifications

Formula Weight 98.15
Chemical Name or Material trans,trans-2,4-Hexadien-1-ol
TSCA TSCA
Name Note stabilized with 0.1% alpha-tocopherol
Assay 98%
Molecular Formula C6H10O
MDL Number MFCD00002925
CAS 17102-64-6
Beilstein 1719718
Synonym 1-Hydroxy-2,4-hexadiene; Sorbyl alcohol
Health Hazard 1 Danger
Boiling Point 79° to 80°C (12mmHg)
Health Hazard 2 GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Melting Point 30° to 31°C
Flash Point 72°C (161°F)
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number MM3325000
Quantity 5g

Applications
Trans,trans-2,4-Hexadien-1-ol is used in the preparation of trans-2,3-methano-4(Z)-hexenol by reacting with diiodomethane. It is used as a flavor and fragrance agent having pineapple odor.

Solubility
Slightly miscible with water.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.