Please login to your online account to display your discounted pricing

Alfa Aesar™ trans,trans-2,4-Decadienal, 90+%, remainder mainly trans,cis isomer

$29.82 - $89.80

Chemical Identifiers

Chemical Name or Material trans,trans-2,4-Decadienal
CAS 25152-84-5
Molecular Formula C10H16O
Formula Weight 152.24
MDL Number MFCD00007007
Beilstein 1704897
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0610303

  SDS
alfa aesar™
L0610303
1g Each for $29.82

AAL0610306

  SDS
alfa aesar™
L0610306
5g Each for $89.80
Description & Specifications

Specifications

Formula Weight 152.24
CAS 25152-84-5
Chemical Name or Material trans,trans-2,4-Decadienal
MDL Number MFCD00007007
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Molecular Formula C10H16O
Health Hazard 1 Warning
Assay ≥90%
Beilstein 1704897
RTECS Number HD3000000
Quantity 1g
Flash Point 101°C (213°F)
Boiling Point 110°C (7mmHg)
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Odor Sweet
Assay Percent Notes remainder mainly trans, cis isomer
TSCA TSCA
Recommended Storage 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

2° to 8°C