missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans,trans-1,5-Bis(4-fluorophenyl)-1,4-pentadien-3-one, TCI America™

$181.81 - $542.38

Chemical Identifiers

CAS 53369-00-9
Molecular Formula C17H12F2O
Molecular Weight (g/mol) 270.279
MDL Number MFCD00298485
InChI Key BNHFGYIPXPENKA-YDWXAUTNSA-N
Synonym trans,trans-Bis(4-fluorobenzal)acetone, trans,trans-Bis(4-fluorobenzylidene)acetone, trans,trans-Bis(4-fluorostyryl) Ketone
PubChem CID 1268251
IUPAC Name (1E,4E)-1,5-bis(4-fluorophenyl)penta-1,4-dien-3-one
SMILES C1=CC(=CC=C1C=CC(=O)C=CC2=CC=C(C=C2)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B22835G View Documents TCI America
B22835G
5g Each for $181.81
 
B228325G View Documents TCI America
B228325G
25g Each for $542.38
 
Specifications

Chemical Identifiers

53369-00-9
270.279
BNHFGYIPXPENKA-YDWXAUTNSA-N
1268251
C1=CC(=CC=C1C=CC(=O)C=CC2=CC=C(C=C2)F)F
C17H12F2O
MFCD00298485
trans,trans-Bis(4-fluorobenzal)acetone, trans,trans-Bis(4-fluorobenzylidene)acetone, trans,trans-Bis(4-fluorostyryl) Ketone
(1E,4E)-1,5-bis(4-fluorophenyl)penta-1,4-dien-3-one

Specifications

Undesignated
5g
C17H12F2O
trans,trans-Bis(4-fluorobenzal)acetone, trans,trans-Bis(4-fluorobenzylidene)acetone, trans,trans-Bis(4-fluorostyryl) Ketone
C1=CC(=CC=C1C=CC(=O)C=CC2=CC=C(C=C2)F)F
270.279
270.28
≥98.0% (HPLC)
154°C
53369-00-9
MFCD00298485
BNHFGYIPXPENKA-YDWXAUTNSA-N
(1E,4E)-1,5-bis(4-fluorophenyl)penta-1,4-dien-3-one
1268251
Crystal-Powder at 20°C
trans,trans-1,5-Bis(4-fluorophenyl)-1,4-pentadien-3-one
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator