missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
trans-N-Fmoc-4-tert-butoxy-L-proline, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6379106
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
trans-N-Fmoc-4-tert-butoxy-L-proline | |
White | |
MFCD00151930 | |
fmoc-4-tert-butoxy-l-proline, 2s-4-tert-butoxy-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carboxylic acid, fmoc-o-tert-butyl-l-hydroxyproline, ksc491o9t, n-alpha-9-fluorenylmethyloxycarbonyl-o-t-butyl-trans-l-hydroxyproline, 2s-1-9h-fluoren-9-ylmethoxycarbonyl-4-2-methylpropan-2-yl oxy pyrrolidine-2-carboxylic acid | |
CC(C)(C)OC1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O | |
409.482 | |
409.47 | |
5 g |
122996-47-8 | |
C24H27NO5 | |
6161467 | |
WPBXBYOKQUEIDW-FXMQYSIJSA-N | |
(2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)-4-[(2-methylpropan-2-yl)oxy]pyrrolidine-2-carboxylic acid | |
6916162 | |
98% | |
Powder |
Chemical Identifiers
122996-47-8 | |
409.482 | |
WPBXBYOKQUEIDW-FXMQYSIJSA-N | |
6916162 | |
CC(C)(C)OC1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O |
C24H27NO5 | |
MFCD00151930 | |
fmoc-4-tert-butoxy-l-proline, 2s-4-tert-butoxy-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carboxylic acid, fmoc-o-tert-butyl-l-hydroxyproline, ksc491o9t, n-alpha-9-fluorenylmethyloxycarbonyl-o-t-butyl-trans-l-hydroxyproline, 2s-1-9h-fluoren-9-ylmethoxycarbonyl-4-2-methylpropan-2-yl oxy pyrrolidine-2-carboxylic acid | |
(2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)-4-[(2-methylpropan-2-yl)oxy]pyrrolidine-2-carboxylic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only