Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-N-Benzyloxycarbonyl-4-tert-butoxy-L-proline, 95%

Chemical Identifiers

Chemical Name or Material trans-N-Benzyloxycarbonyl-4-tert-butoxy-L-proline
CAS 85201-91-8
Formula Weight 321.37
MDL Number MFCD00153336
Beilstein 5088264
Synonym Z-Hyp(tBu)-OH; N-Benzyloxycarbonyl-O-tert-Butyl-L-trans-4-hydroxyproline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6503903

 
alfa aesar™
H6503903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6503906

 
alfa aesar™
H6503906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6503914

 
alfa aesar™
H6503914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 321.37
CAS 85201-91-8
MDL Number MFCD00153336
Physical Form Solid
Chemical Name or Material trans-N-Benzyloxycarbonyl-4-tert-butoxy-L-proline
Assay 95%
Synonym Z-Hyp(tBu)-OH; N-Benzyloxycarbonyl-O-tert-Butyl-L-trans-4-hydroxyproline
Quantity 1g
Storage Note 1 2° to 8°C
Beilstein 5088264
Safety and Handling

2° to 8°C