Please login to your online account to display your discounted pricing

trans-β-Methylstyrene, Stabilized 97%, ACROS Organics™

$300.34 - $300.34

Chemical Identifiers

Chemical Name or Material trans-beta-Methylstyrene
Name Note 97%
CAS 873-66-5
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H10
Linear Formula C6H5CH=CHCH3
Formula Weight 118.18
MDL Number MFCD00009280
Synonym trans-1-Phenyl-1-propene
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC150150010

  SDS
thermo fisher scientific
AC150150010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC150150500

  SDS
thermo fisher scientific
AC150150500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC150150100

  SDS
thermo fisher scientific
150150100
Glass bottle 10g Each for $300.34
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 118.18
Quantity 1g
Color Clear colorless to very faintly yellow
RTECS Number DA8400500
EINECS Number 212-848-0
Density 0.911g/mL
Beilstein 05, 481
Refractive Index 1.5480 to 1.5500 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Stabilizer 0.01% BHT
Infrared Spectrum Authentic
Synonym trans-1-Phenyl-1-propene
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Name Note 97%
Linear Formula C6H5CH=CHCH3
Molecular Formula C9H10
CAS 873-66-5
MDL Number MFCD00009280
Chemical Name or Material trans-beta-Methylstyrene
Assay Percent Range 97%
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.