missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-Methyl crotonate, 96%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 160845000

 View more versions of this product

Catalog No. AC160845000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

trans-Methyl crotonate
-1°C
Glass bottle
0.94
Colorless
98%
95% min. (GC)
CH3CH=CHCO2CH3
02, 410
2-butenoic acid, methyl ester, 2e
MCVVUJPXSBQTRZ-ONEGZZNKSA-N
methyl (E)-but-2-enoate
638132
Liquid
0.9400g/mL
Authentic
1.4230 to 1.4250
120°C
500g
623-43-8
C5H8O2
MFCD00009287
10, 2586
Solubility in water: insoluble
CC=CC(=O)OC
100.12
100.12
96%

Chemical Identifiers

623-43-8
100.12
MCVVUJPXSBQTRZ-ONEGZZNKSA-N
638132
CC=CC(=O)OC
C5H8O2
MFCD00009287
2-butenoic acid, methyl ester, 2e
methyl (E)-but-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear eye protection/face protection.
Wash f

GHS Signal Word: Danger

EINECSNumber : 210-793-7

RTECSNumber : GQ5710000

TSCA : TSCA

SDS Documents