Please login to your online account to display your discounted pricing

trans-Methyl Crotonate 98%, ACROS Organics™

$52.33 - $52.33

Chemical Identifiers

Chemical Name or Material trans-Methyl crotonate
Name Note 98%
CAS 623-43-8
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H8O2
Linear Formula CH3CH=CHCO2CH3
Formula Weight 100.12
MDL Number MFCD00009287
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC160840050

  SDS
thermo fisher scientific
160840050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC160841000

  SDS
thermo fisher scientific
160841000
Glass bottle 100g Each for $52.33

AC160845000

  SDS
thermo fisher scientific
160845000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 100.12
Quantity 5g
Color Clear colorless
TSCA TSCA
RTECS Number GQ5710000
EINECS Number 210-793-7
Density 0.94g/mL
Beilstein 02, 410
Merck Index 10, 2586
Infrared Spectrum Authentic
Refractive Index 1.4230 to 1.4250 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.
Name Note 98%
Linear Formula CH3CH=CHCO2CH3
Assay Percent Range 98%
MDL Number MFCD00009287
Chemical Name or Material trans-Methyl crotonate
Molecular Formula C5H8O2
CAS 623-43-8
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapor.