missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-Indole-3-acrylic acid, 98+%

Manufacturer:  Alfa Aesar™ A1214314

 View more versions of this product

Catalog No. AAA1214314



Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
Specifications

Specifications

trans-Indole-3-acrylic acid
25g
C11H9NO2
6317
PLVPPLCLBIEYEA-AATRIKPKSA-N
(E)-3-(1H-indol-3-yl)prop-2-enoic acid
5375048
≥98%
~185°C (decomposition)
29953-71-7
MFCD00005633
3-indoleacrylic acid, trans-3-indoleacrylic acid, indole-3-acrylic acid, 3-indolylacrylic acid, indole-3beta-acrylic acid, 3-1h-indol-3-yl acrylic acid, indole-3-crylic acid, 2e-3-1h-indol-3-yl prop-2-enoic acid, trans-3-1h-indol-3-yl acrylic acid, indoleacrylic acid van
C1=CC=C2C(=C1)C(=CN2)C=CC(=O)O
187.198
187.2

Chemical Identifiers

29953-71-7
187.198
PLVPPLCLBIEYEA-AATRIKPKSA-N
5375048
C1=CC=C2C(=C1)C(=CN2)C=CC(=O)O
C11H9NO2
MFCD00005633
3-indoleacrylic acid, trans-3-indoleacrylic acid, indole-3-acrylic acid, 3-indolylacrylic acid, indole-3beta-acrylic acid, 3-1h-indol-3-yl acrylic acid, indole-3-crylic acid, 2e-3-1h-indol-3-yl prop-2-enoic acid, trans-3-1h-indol-3-yl acrylic acid, indoleacrylic acid van
(E)-3-(1H-indol-3-yl)prop-2-enoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : NL3680000

Recommended Storage : 2°C to 8°C

SDS Documents