Learn More
trans-Dichlorobis(triphenylphosphine)palladium(II), Pd 14.0% min, Thermo Scientific Chemicals
Hydrogenation, hydrosilation, carbonylation, oxidation, and C-C bond formation
Supplier: Thermo Scientific Chemicals 01049106
Description
As a catalyst of choice in hydrogenation, hydrosilation, carbonylation, oxidation, elimination, reduction, allyl aceate isomerization, dimerization of 1,3-dienes, and C-C bond formation such as Negishi coupling, Suzuki coupling, Kumada coupling, Sonogashira coupling, Heck coupling and Sonogashira-Hagihara coupling reactions. Used in the preparation of tetrakis(triphenylphosphine)palladium, condensed heterocycles such as furoquinolines, diphenylacetylenes, and several other compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
trans-Dichlorobis(triphenylphosphine)palladium(II) | |
∼310°C (decomposition) | |
Odorless | |
PdCl2[P(C6H5)3]2 | |
5 g | |
bis triphenylphosphine palladium ii dichloride, bis triphenylphosphine palladium ii chloride, dichlorobis triphenylphosphine palladium, bis triphenylphosphine palladium chloride, bis triphenylphosphine palladiuml ii dichloride, palladium 2+ ; triphenylphosphane; dichloride, trans-dichlorobis triphenylphosphine palladium ii 1g | |
YNHIGQDRGKUECZ-UHFFFAOYSA-L | |
ethane;methane;palladium(2+);triphenylphosphane;dichloride | |
131664180 | |
Pd 14.0% min. |
13965-03-2 | |
Yellow | |
C36H30Cl2P2Pd | |
MFCD00009593 | |
Hygroscopic | |
Insoluble in water. Soluble in benzene,and toluene. | |
[Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
701.90 | |
701.91 | |
Crystalline |
Chemical Identifiers
13965-03-2 | |
701.90 | |
YNHIGQDRGKUECZ-UHFFFAOYSA-L | |
131664180 | |
[Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
C36H30Cl2P2Pd | |
MFCD00009593 | |
bis triphenylphosphine palladium ii dichloride, bis triphenylphosphine palladium ii chloride, dichlorobis triphenylphosphine palladium, bis triphenylphosphine palladium chloride, bis triphenylphosphine palladiuml ii dichloride, palladium 2+ ; triphenylphosphane; dichloride, trans-dichlorobis triphenylphosphine palladium ii 1g | |
ethane;methane;palladium(2+);triphenylphosphane;dichloride |
Safety and Handling
EINECSNumber : 237-744-2
RTECSNumber : RT3578000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only