Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-Dichlorobis(triethylphosphine)palladium(II)

Coupling of C-C bonds

Chemical Identifiers

Chemical Name or Material trans-Dichlorobis(triethylphosphine)palladium(II)
CAS 28425-04-9
Formula Weight 413.63
MDL Number MFCD00191831
UN Number UN3464
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3982301

  SDS
alfa aesar™
3982301
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3982306

  SDS
alfa aesar™
3982306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material trans-Dichlorobis(triethylphosphine)palladium(II)
CAS 28425-04-9
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331-H319
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.
Formula Weight 413.63
MDL Number MFCD00191831
UN Number UN3464
Solubility Information Soluble in chloroform, toluene and benzene
Quantity 0.25g
Sensitivity Hygroscopic
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331-H319
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye irritation.