Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-Diamminedichloroplatinum(II), Pt 64.5% min

Chemical Identifiers

Chemical Name or Material trans-Diamminedichloroplatinum(II)
CAS 14913-33-8
Linear Formula PtCl2(NH3)2
Formula Weight 300.06
MDL Number MFCD00011623
Synonym Dichlorodiammineplatinum(II); Transplatin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1047201

  SDS
alfa aesar™
1047201
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1047203

  SDS
alfa aesar™
1047203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1047206

  SDS
alfa aesar™
1047206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 300.06
Chemical Name or Material trans-Diamminedichloroplatinum(II)
Physical Form Crystalline
Linear Formula PtCl2(NH3)2
MDL Number MFCD00011623
CAS 14913-33-8
Synonym Dichlorodiammineplatinum(II); Transplatin
Concentration Pt 64.5% min.
Melting Point 340°C (dec.)
Color Yellow
Health Hazard 1 Danger
Quantity 0.25g
Odor Odorless
Health Hazard 2 GHS H Statement
H334-H341-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280h-P262-P305+P351+P338-P308+P313
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
Do not get in eyes, on skin, or on clothing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
RTECS Number TP2455000
Safety and Handling

Danger

GHS P Statement
P261-P280h-P262-P305+P351+P338-P308+P313
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
Do not get in eyes, on skin, or on clothing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or concerned: Get medical advice/attention.

GHS H Statement
H334-H341-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.